Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,4-Dicyano-2-butene
1,4-Dicyano-2-butene 95%
1,4-Dicyano-2-butene 95%
1,4-Dicyano-2-butene 95%
Supplier:
Apollo Scientific
Certificates
Expand image
1,4-Dicyano-2-butene 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
1119-85-3
:
Formula:
C₆H₆N₂
MDL Number:
MFCD00001964
:
106.1252 g/mol
Storage Temperature:
Ambient