Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl (R)-(-)-3-hydroxyisobutyrate
Methyl (R)-(-)-3-hydroxyisobutyrate 97%
Methyl (R)-(-)-3-hydroxyisobutyrate 97%
:
Apollo Scientific
Methyl (R)-(-)-3-hydroxyisobutyrate 97%
:
Apollo Scientific
:
72657-23-9
:
:
C₅H₁₀O₃
:
MFCD00063450
:
118.1311 g/mol
:
Ambient