
Disodium-4-formylbenzene-1,3-disulphonate 98+%
Catalog # APOSOR919217-500G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:310.20 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:33513-44-9
Formula:C₇H₆O₇S₂
MDL Number:MFCD00043913