Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Dichloropyrimidine-4-carbonitrile
2,6-Dichloropyrimidine-4-carbonitrile 97%
2,6-Dichloropyrimidine-4-carbonitrile 97%
2,6-Dichloropyrimidine-4-carbonitrile 97%
Supplier:
Apollo Scientific
Certificates
Expand image
2,6-Dichloropyrimidine-4-carbonitrile 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
26293-93-6
:
Formula:
C₅HCl₂N₃
MDL Number:
MFCD16988336
:
173.9900 g/mol
Storage Temperature:
Ambient