Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dimethyl 3,5-Pyridinedicarboxylate
Dimethyl 3,5-Pyridinedicarboxylate 98%
Dimethyl 3,5-Pyridinedicarboxylate 98%
Dimethyl 3,5-Pyridinedicarboxylate 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Dimethyl 3,5-Pyridinedicarboxylate 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
4591-55-3
:
Formula:
C₉H₉NO₄
MDL Number:
MFCD00518827
:
195.17 g/mol
Storage Temperature:
Ambient