0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
2,4-Dichlorobenzylamine
2,4-Dichlorobenzylamine 98%
2,4-Dichlorobenzylamine 98%
2,4-Dichlorobenzylamine 98%
Supplier:
Apollo Scientific
Certificates
Expand image
2,4-Dichlorobenzylamine 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
95-00-1
:
Formula:
C₇H₇Cl₂N
MDL Number:
MFCD00008109
:
176.04 g/mol
Storage Temperature:
Ambient