Order Entry
Export
ContactUsLinkComponent
Ac-4,5-dehydro-Leu-oh 95%
Ac-4,5-dehydro-Leu-oh 95%
Catalog # APOSOR917451-250MG
Supplier:  Apollo Scientific
CAS Number:  
Ac-4,5-dehydro-Leu-oh 95%
Catalog # APOSOR917451-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :171.196 g/mol
Storage Temperature:Ambient
CAS Number:88547-24-4
Formula:C₈H₁₃NO₃
MDL Number:MFCD00237465