Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,4-Dimethyl-3-hexanol
3,4-Dimethyl-3-hexanol 95%
3,4-Dimethyl-3-hexanol 95%
3,4-Dimethyl-3-hexanol 95%
Supplier:
Apollo Scientific
Certificates
Expand image
3,4-Dimethyl-3-hexanol 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
19550-08-4
:
Formula:
C₈H₁₈O
MDL Number:
MFCD00021820
Storage Temperature:
Ambient