1-Chloro-2,3-dimethoxybenzene 95%
Catalog # APOSOR914653-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Storage Temperature:Ambient
CAS Number:90282-99-8
Formula:C₈H₉ClO₂
MDL Number:MFCD00191399