0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
3-Bromo-4-nitrobenzaldehyde
3-Bromo-4-nitrobenzaldehyde 97%
3-Bromo-4-nitrobenzaldehyde 97%
3-Bromo-4-nitrobenzaldehyde 97%
Supplier:
Apollo Scientific
Certificates
Expand image
3-Bromo-4-nitrobenzaldehyde 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
101682-68-2
:
Formula:
C₇H₄BrNO₃
MDL Number:
MFCD00968939
:
230.02 g/mol
:
GHS07
Storage Temperature:
Ambient