2-Amino-4,5-dimethylthiophene-3-carbonitrile 98%
Catalog # APOSOR914214-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:152.22 g/mol
Storage Temperature:Ambient
CAS Number:4651-94-9
Formula:C₇H₈N₂S
MDL Number:MFCD00720189