2,4-Dimethoxythiophenol 95%
Supplier: Apollo Scientific
Jump to:
About this item
- CAS Number:18906-37-1
- Formula:C₈H₁₀O₂S
- MDL Number:MFCD04973436
- Molecular Weight:170.2288 g/mol
- Storage Temperature:Ambient
3 Options Available Below
Product Details & Documents
- CAS Number:18906-37-1
- Formula:C₈H₁₀O₂S
- MDL Number:MFCD04973436
- Molecular Weight:170.2288 g/mol
- Storage Temperature:Ambient



