Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Ethyl-2,3-dimethylpyrazine
5-Ethyl-2,3-dimethylpyrazine 95%
5-Ethyl-2,3-dimethylpyrazine 95%
5-Ethyl-2,3-dimethylpyrazine 95%
Supplier:
Apollo Scientific
Certificates
Expand image
5-Ethyl-2,3-dimethylpyrazine 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
15707-34-3
:
Formula:
C₈H₁₂N₂
MDL Number:
MFCD04038059
:
136.1900 g/mol
Storage Temperature:
Ambient