Order Entry
Export
ContactUsLinkComponent
2,6-Dichloroquinoxaline 95%
2,6-Dichloroquinoxaline 95%
Catalog # APOSOR912589-5G
Supplier:  Apollo Scientific
CAS Number:  
2,6-Dichloroquinoxaline 95%
Catalog # APOSOR912589-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :199.0368 g/mol
Storage Temperature:Ambient
CAS Number:18671-97-1
Formula:C₈H₄Cl₂N₂
MDL Number:MFCD00839695