Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-4,6-dimethoxypyrimidine
2-Chloro-4,6-dimethoxypyrimidine 98%
2-Chloro-4,6-dimethoxypyrimidine 98%
2-Chloro-4,6-dimethoxypyrimidine 98%
Supplier:
Apollo Scientific
Certificates
Expand image
2-Chloro-4,6-dimethoxypyrimidine 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
13223-25-1
:
Formula:
C₆H₇ClN₂O₂
MDL Number:
MFCD00274530
:
174.58 g/mol
Storage Temperature:
Ambient