Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Temozolomide
Temozolomide 98%
Temozolomide 98%
Irritant
Health
Temozolomide 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Health
Temozolomide 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
85622-93-1
:
Formula:
C₆H₆N₆O₂
MDL Number:
MFCD00866492
:
194.15 g/mol
Melting Pt:
212 °C
:
GHS07
:
GHS08
Storage Temperature:
Refrigerator