Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-(Methylsulphonyl)-1,3-benzothiazol-2-amine
6-(Methylsulphonyl)-1,3-benzothiazol-2-amine 98%
6-(Methylsulphonyl)-1,3-benzothiazol-2-amine 98%
Toxic
6-(Methylsulphonyl)-1,3-benzothiazol-2-amine 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Toxic
6-(Methylsulphonyl)-1,3-benzothiazol-2-amine 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
17557-67-4
:
Formula:
C8H8N2O2S2
Storage Temperature:
Ambient
UN:
2811
ADR:
6.1,III
:
GHS06