Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Methyl-4,6-dimethoxypyrimidine
2-Methyl-4,6-dimethoxypyrimidine 98%
2-Methyl-4,6-dimethoxypyrimidine 98%
2-Methyl-4,6-dimethoxypyrimidine 98%
Supplier:
Apollo Scientific
Certificates
Expand image
2-Methyl-4,6-dimethoxypyrimidine 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
13566-48-8
:
Formula:
C₇H₁₀N₂O₂
MDL Number:
MFCD06657567
:
154.169 g/mol
Storage Temperature:
Ambient