Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 2,3-dihydroxybenzoate
Methyl-2,3-dihydroxybenzoate 98%
Methyl-2,3-dihydroxybenzoate 98%
:
Apollo Scientific
Methyl-2,3-dihydroxybenzoate 98%
:
Apollo Scientific
:
2411-83-8
:
:
C₈H₈O₄
:
MFCD00039520
:
168.1467 g/mol
:
Ambient