Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Methoxy-2,3-dimethylbenzaldehyde
4-Methoxy-2,3-dimethylbenzaldehyde 95%
4-Methoxy-2,3-dimethylbenzaldehyde 95%
4-Methoxy-2,3-dimethylbenzaldehyde 95%
Supplier:
Apollo Scientific
Certificates
Expand image
4-Methoxy-2,3-dimethylbenzaldehyde 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
38998-17-3
:
Formula:
C₁₀H₁₂O₂
MDL Number:
MFCD00003339
:
164.21 g/mol
Storage Temperature:
Ambient