Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
8-Hydroxyimidazo[1,2-a]pyridine-3-carbaldehyde
8-Hydroxyimidazo[1,2-a]pyridine-3-carbaldehyde 96%
8-Hydroxyimidazo[1,2-a]pyridine-3-carbaldehyde 96%
8-Hydroxyimidazo[1,2-a]pyridine-3-carbaldehyde 96%
Supplier:
Apollo Scientific
Certificates
Expand image
8-Hydroxyimidazo[1,2-a]pyridine-3-carbaldehyde 96%
Supplier:
Apollo Scientific
About this item
CAS Number:
1033202-04-8
:
Formula:
C₈H₆N₂O₂
MDL Number:
MFCD10699667
Storage Temperature:
Ambient