Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Methylimidazo[1,2-a]pyridine-3-carbaldehyde
6-Methylimidazo[1,2-a]pyridine-3-carbaldehyde 95%
6-Methylimidazo[1,2-a]pyridine-3-carbaldehyde 95%
6-Methylimidazo[1,2-a]pyridine-3-carbaldehyde 95%
Supplier:
Apollo Scientific
Certificates
Expand image
6-Methylimidazo[1,2-a]pyridine-3-carbaldehyde 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
933752-89-7
:
Formula:
C₉H₈N₂O
MDL Number:
MFCD09994344
:
160.1726 g/mol
Storage Temperature:
Ambient