Order Entry
Export
ContactUsLinkComponent
Ethyl-4,4-imethyl-3-oxovalerate 95%
Irritant
Ethyl-4,4-imethyl-3-oxovalerate 95%
Catalog # APOSOR908242-100G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Ethyl-4,4-imethyl-3-oxovalerate 95%
Catalog # APOSOR908242-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :172.22 g/mol
Boiling Pt:88…90 °C/9.75mmHg
Density:0.967 g/cm³ (25 °C)
Flash Pt:78 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:17094-34-7
Formula:C₉H₁₆O₃
MDL Number:MFCD00042886