Order Entry
Export
ContactUsLinkComponent
Methyl-2,3-dimethoxybenzoate 98%
Methyl-2,3-dimethoxybenzoate 98%
Catalog # APOSOR908171-100G
Supplier:  Apollo Scientific
CAS Number:  
Methyl-2,3-dimethoxybenzoate 98%
Catalog # APOSOR908171-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :196.2 g/mol
Storage Temperature:Ambient
CAS Number:2150-42-7
Formula:C₁₀H₁₂O₄
MDL Number:MFCD00027555