Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 3-methyl-5-nitrobenzoate
Methyl-3-methyl-5-nitrobenzoate 98%
Methyl-3-methyl-5-nitrobenzoate 98%
Methyl-3-methyl-5-nitrobenzoate 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Methyl-3-methyl-5-nitrobenzoate 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
482311-23-9
:
Formula:
C₉H₉NO₄
MDL Number:
MFCD12172988
:
195.1700 g/mol
Storage Temperature:
Ambient