Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,6-Dimethoxynaphthalene
1,6-Dimethoxynaphthalene 98%
1,6-Dimethoxynaphthalene 98%
APOSOR906393-5G
:
Apollo Scientific
:
1,6-Dimethoxynaphthalene 98%
APOSOR906393-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
188.23 g/mol
:
123 °C
:
58…59 °C
:
GHS05
:
Ambient
:
3900-49-0
:
C₁₂H₁₂O₂
:
MFCD00086330