Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Isoxazolo[4,5-b]pyridin-3-amine
Isoxazolo[4,5-b]pyridin-3-amine 98%
Isoxazolo[4,5-b]pyridin-3-amine 98%
Isoxazolo[4,5-b]pyridin-3-amine 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Isoxazolo[4,5-b]pyridin-3-amine 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
114080-93-2
:
Formula:
C₆H₅N₃O
MDL Number:
MFCD13193314
:
135.1200 g/mol
Storage Temperature:
Ambient