1,2-Benzisoxazol-6-ol 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:65685-55-4
:
Formula:C₇H₅NO₂
MDL Number:MFCD09999144
:135.12 g/mol
Boiling Pt:300 °C
Density:1.378 g/cm³ (25 °C)
Flash Pt:135 °C
Storage Temperature:Refrigerator