Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,2-Dimethylpyrrolidine hydrochloride
2,2-Dimethylpyrrolidine hydrochloride 95%
2,2-Dimethylpyrrolidine hydrochloride 95%
2,2-Dimethylpyrrolidine hydrochloride 95%
Supplier:
Apollo Scientific
Certificates
Expand image
2,2-Dimethylpyrrolidine hydrochloride 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
623580-01-8
:
Formula:
C₆H₁₄ClN
MDL Number:
MFCD09759233
:
135.64 g/mol
Storage Temperature:
Ambient