0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
2,2-Dimethyl-1-phenylpropylamine
2,2-Dimethyl-1-phenylpropylamine 98%
2,2-Dimethyl-1-phenylpropylamine 98%
:
Apollo Scientific
2,2-Dimethyl-1-phenylpropylamine 98%
:
Apollo Scientific
:
61501-04-0
:
:
C₁₁H₁₇N
:
MFCD00267378
:
163.26 g/mol
:
GHS07
:
Ambient