Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Dihydroxynaphthalene
2,3-Dihydroxynaphthalene 98%
2,3-Dihydroxynaphthalene 98%
2,3-Dihydroxynaphthalene 98%
Supplier:
Apollo Scientific
Certificates
Expand image
2,3-Dihydroxynaphthalene 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
92-44-4
:
Formula:
C₁₀H₈O₂
MDL Number:
MFCD00004073
:
160.17 g/mol
Melting Pt:
162…164 °C
Flash Pt:
175 °C
Storage Temperature:
Ambient