Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(Methylsulphanyl)[1,3]oxazolo[4,5-b]pyridine
2-(Methylsulphanyl)[1,3]oxazolo[4,5-b]pyridine 95%
2-(Methylsulphanyl)[1,3]oxazolo[4,5-b]pyridine 95%
APOSOR904006-1G
:
Apollo Scientific
:
2-(Methylsulphanyl)[1,3]oxazolo[4,5-b]pyridine 95%
APOSOR904006-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
166.2 g/mol
:
Ambient
:
169205-95-2
:
C₇H₆N₂OS
:
MFCD00973738