Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
8-Chloro-2-methylimidazo[1,2-a]pyrazine
8-Chloro-2-methylimidazo[1,2-a]pyrazine 95%
8-Chloro-2-methylimidazo[1,2-a]pyrazine 95%
8-Chloro-2-methylimidazo[1,2-a]pyrazine 95%
Supplier:
Apollo Scientific
Certificates
Expand image
8-Chloro-2-methylimidazo[1,2-a]pyrazine 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
85333-43-3
:
Formula:
C₇H₆ClN₃
MDL Number:
MFCD09999202
:
167.6 g/mol
Storage Temperature:
Ambient