Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chlorothiazolo[4,5-b]pyridine
2-Chlorothiazolo[4,5-b]pyridine 97%
2-Chlorothiazolo[4,5-b]pyridine 97%
2-Chlorothiazolo[4,5-b]pyridine 97%
Supplier:
Apollo Scientific
Certificates
Expand image
2-Chlorothiazolo[4,5-b]pyridine 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
152170-30-4
:
Formula:
C₆H₃ClN₂S
MDL Number:
MFCD12828130
:
170.62 g/mol
Storage Temperature:
Ambient