Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,6-Dihydroxynaphthalene
1,6-Dihydroxynaphthalene 98%
1,6-Dihydroxynaphthalene 98%
Irritant
1,6-Dihydroxynaphthalene 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1,6-Dihydroxynaphthalene 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
575-44-0
:
Formula:
C₁₀H₈O₂
MDL Number:
MFCD00003981
:
160.17 g/mol
Melting Pt:
135 °C
:
GHS07
Storage Temperature:
Ambient