Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,6-Dimethyl-3-nitropyridine
4,6-Dimethyl-3-nitropyridine 97%
4,6-Dimethyl-3-nitropyridine 97%
:
Apollo Scientific
4,6-Dimethyl-3-nitropyridine 97%
:
Apollo Scientific
:
1074-99-3
:
:
C₇H₈N₂O₂
:
MFCD05982051
:
152.15 g/mol
:
116…118 °C/0.9mmHg
:
20…21 °C
:
GHS07
:
Ambient