Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,6-Dimethyl-3-hydroxypyridine
4,6-Dimethyl-3-hydroxypyridine 97%
4,6-Dimethyl-3-hydroxypyridine 97%
:
Apollo Scientific
4,6-Dimethyl-3-hydroxypyridine 97%
:
Apollo Scientific
:
27296-77-1
:
:
C₇H₉NO
:
MFCD08272851
:
123.16 g/mol
:
GHS07
:
Ambient