Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dihydroxy-6-methoxyquinoline
2,4-Dihydroxy-6-methoxyquinoline 95%
2,4-Dihydroxy-6-methoxyquinoline 95%
2,4-Dihydroxy-6-methoxyquinoline 95%
Supplier:
Apollo Scientific
Certificates
Expand image
2,4-Dihydroxy-6-methoxyquinoline 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
14300-45-9
:
Formula:
C₁₀H₉NO₃
Storage Temperature:
Ambient