Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Dimethyl-4-nitrophenol
2,3-Dimethyl-4-nitrophenol 95%
2,3-Dimethyl-4-nitrophenol 95%
2,3-Dimethyl-4-nitrophenol 95%
Supplier:
Apollo Scientific
Certificates
Expand image
2,3-Dimethyl-4-nitrophenol 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
19499-93-5
:
Formula:
C₈H₉NO₃
MDL Number:
MFCD01861284
:
167.1600 g/mol
Storage Temperature:
Ambient