
1H-Pyrazolo[3,4-b]pyrazin-3-amine
Catalog # APOSOR902364-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:135.1300 g/mol
Melting Pt:244…246 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:81411-64-5
Formula:C₅H₅N₅
MDL Number:MFCD09909663