Order Entry
Export
Orders LinkContactUsLinkComponent
(2-Bromophenoxy)acetic acid 97%

(2-Bromophenoxy)acetic acid 97%

Supplier:   Apollo Scientific
(2-Bromophenoxy)acetic acid 97%
Supplier:  Apollo Scientific
Certificates
(2-Bromophenoxy)acetic acid 97%

5 Options Available Below

Product Details & Documents
  • CAS Number:1879-56-7
  • Formula:(C₆H₄Br)OCH₂COOH
  • MDL Number:MFCD00234407
  • Molecular Weight:231.05 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 31,90 €
    31,90 €
    1 * 1 g
       31,90 €

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 105,00 €
    105,00 €
    1 * 5 g
       105,00 €

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 207,00 €
    207,00 €
    1 * 25 g
       207,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 805,00 €
    805,00 €
    1 * 100 g
       805,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 3 860,00 €
    3 860,00 €
    1 * 500 g
       3 860,00 €

    Specifications:
    New Product
    New Product