Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,2-Dimethylmorpholine
2,2-Dimethylmorpholine 95%
2,2-Dimethylmorpholine 95%
2,2-Dimethylmorpholine 95%
Supplier:
Apollo Scientific
Certificates
Expand image
2,2-Dimethylmorpholine 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
147688-58-2
:
Formula:
C₆H₁₃NO
MDL Number:
MFCD09701959
:
115.18 g/mol
Storage Temperature:
Ambient