

α,α'-Dibromo-o-xylene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:91-13-4
:
Formula:C₈H₈Br₂
MDL Number:MFCD00000175
:263.96 g/mol
Melting Pt:91…96 °C
Density:1.96 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:202-042-7
UN:3261
ADR:8,II
:GHS05
:GHS07