Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 5-bromovalerate
Methyl-5-bromovalerate
Methyl-5-bromovalerate
Methyl-5-bromovalerate
Supplier:
Apollo Scientific
Certificates
Expand image
Methyl-5-bromovalerate
Supplier:
Apollo Scientific
About this item
CAS Number:
5454-83-1
:
Formula:
BrH₂C(CH₂)₃COOCH₃
MDL Number:
MFCD00000265
:
195.06 g/mol
Density:
1.363 g/cm³ (25 °C)
Flash Pt:
99 °C
Storage Temperature:
Ambient