

1,2-Dichlorobenzene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:95-50-1
:
Formula:C₆H₄Cl₂
MDL Number:MFCD00000535
:147 g/mol
Boiling Pt:178…180 °C
Melting Pt:–18…–17 °C
Density:1.306 g/cm³ (25 °C)
Flash Pt:66 °C
Storage Temperature:Ambient
EINECS:202-425-9
UN:1591
ADR:6.1,III
:GHS07
:GHS09