Order Entry
Export
Orders LinkContactUsLinkComponent
2,6-Dimethylbenzaldehyde

2,6-Dimethylbenzaldehyde

Irritant
Supplier:   Apollo Scientific
2,6-Dimethylbenzaldehyde
Supplier:  Apollo Scientific
Certificates
2,6-Dimethylbenzaldehyde

3 Options Available Below

Product Details & Documents
Synonyms:  2-Formyl-m-xylene
  • CAS Number:1123-56-4
  • Formula:C₉H₁₀O
  • MDL Number:MFCD00128003
  • Molecular Weight:134.18 g/mol
  • Boiling Pt:230 °C
  • Melting Pt:27…30 °C
  • Flash Pt:96 °C
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 21,80 €
    21,80 €
    1 * 5 g
       21,80 €

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 60,80 €
    60,80 €
    1 * 25 g
       60,80 €

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 170,00 €
    170,00 €
    1 * 100 g
       170,00 €