Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Dibromo-2-chloropyridine
3,5-Dibromo-2-chloropyridine 97%
3,5-Dibromo-2-chloropyridine 97%
Irritant
3,5-Dibromo-2-chloropyridine 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3,5-Dibromo-2-chloropyridine 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
40360-47-2
:
Formula:
C₅H₂Br₂ClN
MDL Number:
MFCD00233991
:
271.34 g/mol
:
GHS07
Storage Temperature:
Ambient