Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,3-Dichloro-6-methoxyisoquinoline
1,3-Dichloro-6-methoxyisoquinoline 98%
1,3-Dichloro-6-methoxyisoquinoline 98%
:
Apollo Scientific
1,3-Dichloro-6-methoxyisoquinoline 98%
:
Apollo Scientific
:
24623-39-0
:
:
C₁₀H₇Cl₂NO
:
MFCD09258765
:
228.747 g/mol
:
GHS07
:
Refrigerator