Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-[(Trimethylsilyl)ethynyl]benzoic acid
4-[(Trimethylsilyl)ethynyl]benzoic acid
4-[(Trimethylsilyl)ethynyl]benzoic acid
Irritant
4-[(Trimethylsilyl)ethynyl]benzoic acid
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-[(Trimethylsilyl)ethynyl]benzoic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
16116-80-6
:
Formula:
C₁₂H₁₄O₂Si
MDL Number:
MFCD06203324
:
218.33 g/mol
Melting Pt:
177…178 °C
:
GHS07
Storage Temperature:
Refrigerator