Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,5-Dichloronicotinic acid
4,5-Dichloronicotinic acid
4,5-Dichloronicotinic acid
Irritant
4,5-Dichloronicotinic acid
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4,5-Dichloronicotinic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
473837-10-4
:
Formula:
C₆H₃Cl₂NO₂
MDL Number:
MFCD01861987
:
192 g/mol
:
GHS07
Storage Temperature:
Ambient